Learn More
Sino Biological Anti-LDLR/LDL Receptor Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10231MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LDLR / LDL R / LDL Receptor (rh LDLR / LDL R / LDL Receptor; Catalog#10231-H08H; NP_000518.1; Met1-Arg788). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8868