Learn More
Sino Biological Anti-LDLR/LDL Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10231T50100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LDLR/LDL Receptor (rh LDLR/LDL Receptor; Catalog#10231-H08H; NP_000518.1; Met1-Arg788). LDLR/LDL Receptor specific IgG was purified by Human LDLR/LDL Receptor affinity chromatography.
Catalog No. 50-255-5420