Learn More
Sino Biological Recombinant Human LDLR/LDL Receptor Protein (mFc Tag)
Supplier:  Sino Biological 10231H05H100
Encompass_Preferred
A DNA sequence encoding the human LDLR (NP_000518.1) (Met1-Arg788) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-241-1181