Learn More
Sino Biological Recombinant Human LDLR/LDL Receptor Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10231H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human LDLR (NP_000518.1) precursor (Met1-Arg788) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5421