Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LDOC1 Full-length ORF (NP_036449.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89950520 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-520 25 μg
89-950-519 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-950-520 Supplier Abnova™ Supplier No. H00023641P01L
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers. [provided by RefSeq]

Sequence: MVDELVLLLHALLMRHRALSIENSQLMEQLRLLVCERASLLRQVRPPSCPVPFPETFNGESSRLPEFIVQTASYMLVNENRFCNDAMKVAFLISLLTGEAEEWVVPYIEMDSPILGDYRAFLDEMKQCFGWDDDEDDDDEEEEDDY

Specifications

Accession Number NP_036449.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23641
Molecular Weight (g/mol) 43.4kDa
Name LDOC1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MVDELVLLLHALLMRHRALSIENSQLMEQLRLLVCERASLLRQVRPPSCPVPFPETFNGESSRLPEFIVQTASYMLVNENRFCNDAMKVAFLISLLTGEAEEWVVPYIEMDSPILGDYRAFLDEMKQCFGWDDDEDDDDEEEEDDY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BCUR1/Mar7/Mart7
Common Name LDOC1
Gene Symbol LDOC1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.