Learn More
Biomatik Corporation Human Lecithin Cholesterol Acyltransferase (LCAT) CLIA Kit
Supplier:  Biomatik Corporation EKU09980
Encompass_Preferred
Human Lecithin Cholesterol Acyltransferase (LCAT) CLIA Kit for detecting LCAT in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.21-150ng/ml. Sensitivity: 0.08ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04180. Gene ID: 3931. Target Synonyms: phosphatidylcholine-sterol O-acyltransferase; Phospholipid-cholesterol acyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6782