Learn More
BIOHIPPO Human LECT2 Microsample
SDP
Catalog No. 5000314838
Encompass
Human LECT2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LECT2 in Serum plasma and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 31 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LECT2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LECT2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LECT2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14838 Supplier BIOHIPPO Supplier No. BHE15211624
Human LECT2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LECT2 in Serum plasma and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 31 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LECT2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LECT2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LECT2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color