Learn More
Biomatik Corporation Human Lectin, Mannose Binding 2 (LMAN2) ELISA Kit
Supplier:  Biomatik Corporation EKU05580
Encompass_Preferred
Human Lectin, Mannose Binding 2 (LMAN2) ELISA Kit for detecting LMAN2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12907. Gene ID: 10960. Target Synonyms: GP36B; VIP36; C5orf8; Vesicular integral-membrane protein VIP36; Glycoprotein GP36b; Vesicular integral-membrane protein 36. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6507