Learn More
Biomatik Corporation Human Left/Right Determination Factor 1 (LEFTY1) CLIA Kit
Supplier:  Biomatik Corporation EKU09828
Encompass_Preferred
Human Left/Right Determination Factor 1 (LEFTY1) CLIA Kit for detecting LEFTY1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.69-500pg/ml. Sensitivity: 0.25pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O75610. Gene ID: 10637. Target Synonyms: LEFTY-B; LEFTYB; LEFT-B; LEFTB; Left Right Determination Factor B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7746