Learn More
Biomatik Corporation Human Lens Epithelium Derived Growth Factor (LEDGF) ELISA Kit
Supplier:  Biomatik Corporation EKU12437
Encompass_Preferred
Human Lens Epithelium Derived Growth Factor (LEDGF) ELISA Kit for detecting LEDGF in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75475. Gene ID: 11168. Target Synonyms: PSIP1; p75; p52; DFS70; PAIP; PSIP2; PC4 And SFRS1 Interacting Protein 1; CLL-associated antigen KW-7; Dense fine speckles 70 kDa protein; Transcriptional coactivator p75/p52. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5290