Learn More
Biomatik Corporation Human Leucine Aminopeptidase (LAP) ELISA Kit
Supplier:  Biomatik Corporation EKU05601
Encompass_Preferred
Human Leucine Aminopeptidase (LAP) ELISA Kit for detecting LAP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28838. Gene ID: 51056. Target Synonyms: LAPEP, LAP3; PEPS; Peptidase S; Leucyl Aminopeptidase; Cytosol aminopeptidase; Proline aminopeptidase; Prolyl aminopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8894