Learn More
Biomatik Corporation Human Leucine Rich Repeat Containing Protein 3C (LRRC3C) CLIA Kit
Supplier:  Biomatik Corporation EKU10018
Encompass_Preferred
Human Leucine Rich Repeat Containing Protein 3C (LRRC3C) CLIA Kit for detecting LRRC3C in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: A6NJW4. Gene ID: 100505591. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7153