Learn More
Biomatik Corporation Human Leucine Rich Repeat LGI Family, Member 3 (LGI3) ELISA Kit
Supplier:  Biomatik Corporation EKU05612
Encompass_Preferred
Human Leucine Rich Repeat LGI Family, Member 3 (LGI3) ELISA Kit for detecting LGI3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N145. Gene ID: 203190. Target Synonyms: LGIL4; LGI1-like protein 4; Leucine-rich glioma-inactivated protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8893