Learn More
American Research Products Inc Human leucocyte antigen B27 H
Catalog No. 5000326156
Encompass
Human leucocyte antigen B27 HLA-B27 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum urine cerebrospinal fluid (CSF) Method Sandwich
Catalog No. 50-003-26156 Supplier American Research Products Inc Supplier No. CSBE08607H
Human leucocyte antigen B27 HLA-B27 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum urine cerebrospinal fluid (CSF) Method Sandwich