Learn More
Biomatik Corporation Human Leukocyte Cell Derived Chemotaxin 1 (LECT1) ELISA Kit
Supplier:  Biomatik Corporation EKU05621
Encompass_Preferred
Human Leukocyte Cell Derived Chemotaxin 1 (LECT1) ELISA Kit for detecting LECT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75829. Gene ID: 11061. Target Synonyms: BRICD3; CHM-I; CHM1; CH-SP; Chondromodulin-1; Chondrosurfactant protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5674