Learn More
American Research Products Inc Human Leukotriene B4 LT B4 EL
Catalog No. 5000328889
Encompass
Human Leukotriene B4 LT-B4 ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 31 ng/mL Sample serum urine cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28889 Supplier American Research Products Inc Supplier No. CSBE08033H
Human Leukotriene B4 LT-B4 ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 31 ng/mL Sample serum urine cell culture supernates tissue homogenates Method Sandwich