Learn More
Bioworld HUMAN LEUPROLIDE
Supplier:  Bioworld 220604071
Encompass_Preferred
Human Leuprolide (Leuprorelin) Source: Description : Synthetic Human Leuprolide is a single, non-glycosylated, polypeptide chain containing 9 amino acids and having a molecular mass of 1209 Dalton. Physical Appearance: Sterile Filt...
Catalog No. 50-253-503