Learn More
BIOHIPPO Human LFA3 Microsample
SDP
Catalog No. 5000315644
Encompass
Human LFA3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LFA3 in Serum plasma cell lysatesand other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LFA3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LFA3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LFA3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15644 Supplier BIOHIPPO Supplier No. BHE15212592
Human LFA3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LFA3 in Serum plasma cell lysatesand other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LFA3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LFA3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LFA3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color