Learn More
Description
A positive reactivity in plasma B-cells is indicated by cytoplasmic staining. In lymphoid tissue, the normal kappa-to-lambda ratio is roughly 2:1, an indication for monoclonality is given if the kappa-to-lambda ratio is greater than 3:1 or less than 0.3:1. Visualization of signals should be performed at least at 100-fold magnification resulting in easily visible signals. Do not evaluate areas of necrosis, overlapping nuclei, over-digested nuclei, and nuclei with weak signal intensity.
Specifications
Specifications
| Content And Storage | Ready-to-use. No reconstitution, mixing, or dilution required. Bring to room temperature and mix briefly before use. Typically used with Human Ig-lambda CISH Probe |
| For Use With (Application) | CISH-P |
| Includes | Reagent Provided: Digoxigenin-labeled oligonucleotides targeting mRNA sequences encoding Ig-kappa light chain constant regions. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.