Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human lg-kappa/lg-lambda CISH Permanent Kit

Catalog No. 89335587 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-587 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-587 Supplier Abnova™ Supplier No. KA5368
Add to Cart
Add to Cart

Human lg-kappa/lg-lambda CISH Permanent Kit is intended to be used for the simultaneous qualitative detection of human Ig-kappa (κ) and Ig-lambda (λ) light chain mRNA in formalin-fixed, paraffin-embedded specimens such as lymphomas by chromogenic in situ hybridization (CISH).

Specifications

Content And Storage Store at 2 to 8°C in an upright position. Return to storage conditions immediately after use.
For Use With (Application) CISH-P
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.