Learn More
Abnova™ Human lg-kappa/lg-lambda CISH Probe
Catalog No. 89335582 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-582 CISH-P
1 options
Catalog No. 89-335-582 Supplier Abnova™ Supplier No. CG0031
Add to Cart
Add to Cart
Human lg-kappa/lg-lambda CISH Probe is designed for the simultaneous qualitative detection of human Ig-kappa and Ig-lambda light chain mRNA in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
The probe is intended to be used in combination with the Human Ig-kappa/Ig-lambda CISH Permanent Kit, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
A positive reactivity in plasma B-cells is indicated by cytoplasmic staining. In lymphoid tissue, the normal kappa-to-lambda ratio is roughly 2:1, an indication for monoclonality is given if the kappa-to-lambda ratio is greater than 3:1 or less than 0.3:1. Visualization of signals should be performed at least at 100-fold magnification resulting in easily visible signals. Do not evaluate areas of necrosis, overlapping nuclei, over-digested nuclei, and nuclei with weak signal intensity.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (55°C) and mix briefly before use.
For Use With (Application) CISH-P
Includes Reagent Provided:
1.Digoxigenin-labeled oligonucleotides targeting mRNA sequences encoding Ig-kappa light chain constant regions.
2. Biotin-labeled oligonucleotides targeting mRNA sequences encoding Ig-lambda light chain constant regions.