Learn More
Sino Biological Recombinant Human Galectin-1/LGALS1 Protein (His Tag)
Supplier:  Sino Biological 10290H07E100
Encompass_Preferred
A DNA sequence encoding the Human LGALS1 (NP_002296.1) (Gln56-Leu279) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-4351