Learn More
Sino Biological Recombinant Anti-Galectin-3/LGALS3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10289R17920
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human LGALS3 (rh LGALS3; Catalog#10289-HNAE; P17931; Met1-Ile250).
Catalog No. 50-241-1672