Learn More
Sino Biological Recombinant Human Galectin-3/LGALS3 Protein (GST Tag), HPLC-verified
Supplier:  Sino Biological 10289H09E1
Encompass_Preferred
A DNA sequence encoding the Human LGALS3 (NP_002297.2) (Ala2-Ile250) was fused with the GST tag at the N-terminus.
Catalog No. 50-255-5394