Learn More
Sino Biological Anti-Galectin-9/LGALS9 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11147MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Galectin 9 (rh Galectin 9; Catalog#11147-H01H; NP_002299.2; Ala2-Thr323). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4814