Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LGP2 Synthetic Peptide

Catalog No. PEP0552 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0552 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0552 Supplier Invitrogen™ Supplier No. PEP0552
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 11 amino acids near the center of human LGP2. PEP-0552 can be used as a blocking peptide with polyclonal antibody PA5-20435.

Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 79132
Species Human
Purification Method purified
Accession Number Q96C10
Common Name LGP2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias B430001I08Rik; D11LGP2; D11lgp2e; DEXH (Asp-Glu-X-His) box polypeptide 58; DEXH-box helicase 58; DHX58; Lgp2; LPG2; ortholog of mouse D11lgp2; probable ATP-dependent helicase LGP2; Probable ATP-dependent RNA helicase DHX58; protein D11Lgp2; Protein D11Lgp2 homolog; RGD1310093; RIG-I-like receptor 3; RIG-I-like receptor Lgp2; RLR; RLR-3; RNA helicase LGP2
Gene Symbol DHX58
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.