Learn More
ACROBiosystems Human LGR4 / GPR48 protein, His Tag
Catalog No. 502054894
Encompass_Preferred
Recombinant Protein;1MG;Human LGR4, His Tag (LG4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 25 -Thr 544 (Accession # Q9BXB1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.0 kDa.
Catalog No. 50-205-4894 Supplier ACROBiosystems Supplier No. LG4H52H31MG
Recombinant Protein;1MG;Human LGR4, His Tag (LG4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 25 -Thr 544 (Accession # Q9BXB1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.0 kDa.