Learn More
Sino Biological Recombinant Human LGR5 Protein (VLP, Flag & His-Strep Tag)
Catalog No. 502993177
Encompass_Preferred
A DNA sequence encoding the Human LGR5 -VLP (O75473) (Gly22-Ser844) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.
Catalog No. 50-299-3177 Supplier Sino Biological Supplier No. 30115H82H20
A DNA sequence encoding the Human LGR5 -VLP (O75473) (Gly22-Ser844) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.