Learn More
ACROBiosystems ACROBiosystems Human LIF R / CD118 Protein, Fc Tag
Catalog No. 502106710
Encompass_Preferred
Recombinant Protein;50UG;Human LIF R, Fc Tag (LIR-H4252) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - Ser 833 (Accession # P42702-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 115.5 kDa. The protein migrates as 140-190 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human LIF, Tag Free (Cat. No. LIF-H521b) at 0.2 μg/mL (100 μL/well) can bind Human LIF R, Fc Tag (Cat. No. LIR-H4252) with a linear range of 4-125 ng/mL (QC tested).
Catalog No. 50-210-6710 Supplier ACROBiosystems Supplier No. LIRH425250UG
Add to Cart
Add to Cart
Recombinant Protein;50UG;Human LIF R, Fc Tag (LIR-H4252) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - Ser 833 (Accession # P42702-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 115.5 kDa. The protein migrates as 140-190 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human LIF, Tag Free (Cat. No. LIF-H521b) at 0.2 μg/mL (100 μL/well) can bind Human LIF R, Fc Tag (Cat. No. LIR-H4252) with a linear range of 4-125 ng/mL (QC tested).