Learn More
Sino Biological Anti-LIFR Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10628MM0720
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LIFR / CD118 (rh LIFR; Catalog#10628-H08H; NP_001121143.1; Met 1-Ser 833). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-0659