Learn More
Sino Biological Anti-LIFR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10628RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LIFR / CD118 (rh LIFR; Catalog#10628-H08H; NP_001121143.1; Met 1-Ser 833). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2248