Learn More
Sino Biological Recombinant Human LIFR Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10628H08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human LIFR (NP_001121143.1) (Met 1-Ser 833) with a C-terminal polyhistidine tag was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5233