Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human LIGHT Recombinant Protein, PeproTech®

Catalog No. 31009B15UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 μg
3 μg
7 x 15 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31009B15UG 15 μg
31-009-B100 7 x 15 μg
310-09B-3UG 3 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 31009B15UG Supplier Gibco™ Supplier No. 31009B15UG
  • $224.00
    $190.65 / Each of 1

    Save $33.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

310-09B-100UG will be provided as 15 μg (310-09B-15UG) vials. 310-09B-1MG will be provided as 15 μg (310-09B-15UG) vials. 310-09B-250UG will be provided as 15 μg (310-09B-15UG) vials. 310-09B-500UG will be provided as 15 μg (310-09B-15UG) vials. Recombinant Human LIGHT has a calculated mass of 19.3 kDa, containing 177 amino acid residues. Due to glycosylation, LIGHT migrates between 20.0-22.5 kDa by SDS-PAGE under non-reducing conditions. *Human LIGHT(Catalog Number 310-09B) has replaced Human LIGHT(Catalog Number 310-09) This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43557
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 8740
Molecular Weight (g/mol) 19.3 kDa
Name Human LIGHT
Purification Method Purified
Quantity 15 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias CD258; Herpes virus entry mediator ligand; herpesvirus entry mediator ligand; HVEML; HVEM-L; Light; LTg; Ly113; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tnfsf14; TR2; tumor necrosis factor (ligand) superfamily, member 14; tumor necrosis factor ligand 1D; tumor necrosis factor ligand superfamily member 14; Tumor necrosis factor ligand superfamily member 14, membrane form; Tumor necrosis factor ligand superfamily member 14, soluble form; tumor necrosis factor superfamily member 14; UNQ391/PRO726
Common Name LIGHT
Gene Symbol TNFSF14
Biological Activity Determined by its ability to induce IL-8 secretion in human BEAS-2 B cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence RLGEMVTRLP DGPAGSWEQL IQERRSHEVN PAAHLTGANS SLTGSGGPLL WETQLGLAFL RGLSYHDGAL VVTKAGYYYI YSKVQLGGVG CPLGLASTIT HGLYKRTPRY PEELELLVSQ QSPCGRATSS SRVWWDSSFL GGVVHLEAGE KVVVRVLDER LVRLRDGTRS YFGAFMV
Content And Storage -20°C
Activity Determined by its ability to induce IL-8 secretion in human BEAS-2B cells.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.