Learn More
Sino Biological Recombinant Human Thy1/CD90 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16897H08H500
Encompass_Preferred
A DNA sequence encoding the human THY1 (NP_006279.2) (Met1-Cys130) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-222-3685