Learn More
Sino Biological Recombinant Human LILRA1/LIR-6/CD85i Protein (hFc Tag)
Supplier:  Sino Biological 17220H02H100
Encompass_Preferred
A DNA sequence encoding the human LILRA1 (NP_006854.1) (Met1-Asn461) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4011