Learn More
Sino Biological Recombinant Human ILT1/LILRA2/LIR-7 Protein (hFc Tag)
Supplier:  Sino Biological 30010H02H100
Encompass_Preferred
A DNA sequence encoding the human LILRA2 (NP_001124389.2) (Met1-Asn449) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3812