Learn More
Sino Biological Anti-LILRA3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13549MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ILT6 / LILRA3 (rh ILT6 / LILRA3; Catalog#13549-H08H; AAH28208.1; Met1-Glu439). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8940