Learn More
Sino Biological Human LILRA3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13549T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ILT6 / LILRA3 (rh ILT6 / LILRA3; Catalog#13549-H08H; AAH28208.1; Met1-Glu439). ILT6 / LILRA3 specific IgG was purified by Human ILT6 / LILRA3 affinity chromatography.
Catalog No. 50-237-1436