Learn More
Sino Biological Anti-ILT11/LILRA5/LIR-9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 16059RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LILRA5 (rh LILRA5; Catalog#16059-H08H; NP_067073.1; Met1-Arg268). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1256