Learn More
Sino Biological Human LILRA5 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 16059T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LILRA5 (rh LILRA5; Catalog#16059-H08H; NP_067073.1; Met1-Arg268). LILRA5 specific IgG was purified by Human LILRA5 affinity chromatography.
Catalog No. 50-237-2367