Learn More
Sino Biological Recombinant Human ILT11/LILRA5/LIR-9 Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 16059H27HB20
Encompass_Preferred
A DNA sequence encoding the human LILRA5 (NP_067073.1) (Met1-Arg268) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4065