Learn More
Sino Biological Recombinant Human ILT11/LILRA5/LIR-9 Protein (hFc Tag) 1
Supplier:  Sino Biological 16059H02H1
Encompass_Preferred
A DNA sequence encoding the human LILRA5 (NP_067073.1) (Met1-Arg268) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6870