Learn More
Sino Biological Recombinant Human ILT-8/LILRA6 Protein (His Tag)
Supplier:  Sino Biological 29813H08H50
Encompass_Preferred
A DNA sequence encoding the human LILRA6 (Q6PI73-1) (Met1-Asn447) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-255-3817