Learn More
ACROBiosystems Human LILRB1 / CD85j / ILT2 (23-431) Protein, His Tag (MALS verified)
Catalog No. 502331393
Encompass_Preferred
Human LILRB1 (23-431) Protein, His Tag (CDJ-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Thr 431 (Accession # Q8NHL6-1).
Catalog No. 50-233-1393 Supplier ACROBiosystems Supplier No. CDJH52H6100UG
Human LILRB1 (23-431) Protein, His Tag (CDJ-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Thr 431 (Accession # Q8NHL6-1).