Learn More
Sino Biological Recombinant Human ILT2/LILRB1 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 16014H02H500
Encompass_Preferred
A DNA sequence encoding the human LILRB1 (ADJ55949.1) (Met1-His458) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-222-3672