Learn More
Sino Biological Human LILRB1 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 16014MM06P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD85 / ILT2 / ILR1 (rh CD85 / ILT2 / ILR1; Catalog#16014-H08H; ADJ55949.1; Met1-His458) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-2672