Learn More
Sino Biological Anti-ILT2/LILRB1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 16014MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD85 / ILT2 / ILR1 / LILRB1 (rh CD85 / ILT2 / ILR1 / LILRB1; Catalog#16014-H08H; ADJ55949.1; Met1-His458). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1096