Learn More
Sino Biological Human LILRB1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 16014T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD85 / ILT2 / ILR1 / LILRB1 (rh CD85 / ILT2 / ILR1 / LILRB1; Catalog#16014-H08H; ADJ55949.1; Met1-His458). CD85 / ILT2 / ILR1 / LILRB1 specific IgG was purified by Human CD85 / ILT2 / ILR1 / LILRB1 affinity chromatography.
Catalog No. 50-237-1567