Learn More
Sino Biological Recombinant Human ILT2/LILRB1 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 16014H49HB20
Encompass_Preferred
A DNA sequence encoding the Human LILRB1 (ADJ55949.1) (Met1-His458) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1467