Learn More
Abcam Recombinant Human LILRB1 Protein, 50UG
Supplier:  Abcam AB29010650UG
Encompass_Preferred
Recombinant Human LILRB1 Protein is a Human Fragment protein, in the 24 to 458 aa range, expressed in HEK 293, with >=95% purity, <= 0.005 EU/µg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: CD85j, ILT2, LIR1, MIR7, LILRB1, Leukocyte immunoglobulin-like receptor subfamily B member 1, LIR-1, Leukocyte immunoglobulin-like receptor 1, CD85 antigen-like family member J, Immunoglobulin-like transcript 2, Monocyte/macrophage immunoglobulin-like receptor 7, ILT-2, MIR-7
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2742177